Learn More
Medchemexpress LLC 3-(1H-indol-3-yl)propanoic-2,2-d2 acid | 2469257-98-3 | 99.5% | 191.22 | C11H9D2NO2 | 10 MG
SDP
Catalog No. 5000302537
Encompass_Preferred
3-Indolepropionic acid-d2 is the deuterium-labeled isotopologue of 3-indolepropionic acid used as an analytical standard and tracer in biochemical and mass spectrometry studies. It provides a mass-shifted internal standard for quantitative assays and metabolic research while retaining the chemical behavior of the parent compound.
  • Deuterium label enables accurate mass spectrometry quantitation.
  • High chemical purity supports reliable analytical results.
  • Solid powder form suitable for storage and reconstitution.
  • Useful as an internal standard or tracer in metabolic studies.
  • Relevant for studies of antioxidant activity and neurobiology.
Catalog No. 50-003-02537 Supplier Medchemexpress LLC Supplier No. HYW015229S10MG
3-Indolepropionic acid-d2 is the deuterium-labeled isotopologue of 3-indolepropionic acid used as an analytical standard and tracer in biochemical and mass spectrometry studies. It provides a mass-shifted internal standard for quantitative assays and metabolic research while retaining the chemical behavior of the parent compound.
  • Deuterium label enables accurate mass spectrometry quantitation.
  • High chemical purity supports reliable analytical results.
  • Solid powder form suitable for storage and reconstitution.
  • Useful as an internal standard or tracer in metabolic studies.
  • Relevant for studies of antioxidant activity and neurobiology.