Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1H-Pyrazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. CC56114DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC56114DE 5 g
CC56114CB 250 mg
CC56114DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC56114DE Supplier Thermo Scientific Chemicals Supplier No. CC56114DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 892502-09-9
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
InChI Key JGTYTUGTENJXBY-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-1-ylmethyl aniline, 3-1h-pyrazol-1-yl methyl aniline, 3-pyrazol-1-ylmethyl-phenylamine, 3-pyrazol-1-ylmethyl aniline, 3-1-pyrazolyl methyl aniline, 3-1h-pyrazol-1-ylmethyl phenyl amine, 3-pyrazolylmethyl phenylamine
PubChem CID 6485349
IUPAC Name 3-(pyrazol-1-ylmethyl)aniline
SMILES C1=CC(=CC(=C1)N)CN2C=CC=N2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.