Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(1H-Pyrazol-3-yl)benzoic acid, 97%, Thermo Scientific™

Catalog No. CC31701CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC31701CB 250 mg Amber Glass Bottle
CC31701DA 1 g Amber Glass Bottle
CC31701DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC31701CB Supplier Thermo Scientific Chemicals Supplier No. CC31701CB
Add to Cart

Chemical Identifiers

CAS 850375-11-0
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.19
MDL Number MFCD03407956
InChI Key RXZRBZWATRFHCS-UHFFFAOYSA-N
Synonym 3-1h-pyrazol-5-yl benzoic acid, 3-1h-pyrazol-3-yl benzoic acid, 3-2h-pyrazol-3-yl benzoic acid, benzoic acid, 3-1h-pyrazol-3-yl, 5-3-carboxyphenyl-1h-pyrazole, pubchem22748, buttpark 44\01-87, d0tx9p, 3-pyrazol-3-ylbenzoic acid, 3-pyrazol-5-ylbenzoic acid
PubChem CID 2795542
IUPAC Name 3-(1H-pyrazol-5-yl)benzoic acid
SMILES OC(=O)C1=CC=CC(=C1)C1=CC=NN1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 3-(1H-Pyrazol-3-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.