Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[[(1R,2S)-1-hydroxy-1-phenylpropan-2-yl]amino]-1-(3-methoxyphenyl)propan-1-one;hydrochloride | 16777-42-7 | 99.9% | 349.85 g·mol⁻¹ | C19H24ClNO3 | 5 MG
SDP

Catalog No. 5000243265
Encompass_Preferred

Oxyfedrine hydrochloride is a vasodilator and an orally active β-adrenoreceptor agonist used in cardiovascular research. It decreases the tonicity of coronary vessels and has documented in vitro and in vivo effects on aldehyde dehydrogenase activity, myogenic activity, and blood viscosity.

  • High purity (99.92%) suitable for research applications.
  • Orally active β-adrenoreceptor agonist with vasodilatory activity.
  • Molecular formula C19H24ClNO3; molecular weight 349.85 g·mol⁻¹.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in DMSO (175 mg/mL); ultrasonic and newly opened DMSO recommended.
  • Certificate of analysis available to support quality control.

Catalog No. 50-002-43265 Supplier Medchemexpress LLC Supplier No. HY1083005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxyfedrine hydrochloride is a vasodilator and an orally active β-adrenoreceptor agonist used in cardiovascular research. It decreases the tonicity of coronary vessels and has documented in vitro and in vivo effects on aldehyde dehydrogenase activity, myogenic activity, and blood viscosity.

  • High purity (99.92%) suitable for research applications.
  • Orally active β-adrenoreceptor agonist with vasodilatory activity.
  • Molecular formula C19H24ClNO3; molecular weight 349.85 g·mol⁻¹.
  • Solid, white to off-white appearance for easy handling.
  • Soluble in DMSO (175 mg/mL); ultrasonic and newly opened DMSO recommended.
  • Certificate of analysis available to support quality control.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.