Learn More
Medchemexpress LLC 3-[(1S)-1-(dimethylamino)ethyl]phenol | 139306-10-8 | MFCD06656491 | 99.0% | 165.23 g·mol⁻1 | C10H15NO | 25 G
SDP
Catalog No. 5000303356
Encompass_Preferred
Rivastigmine metabolite (NAP 226-90), 3-[(1S)-1-(dimethylamino)ethyl]phenol, is the primary active metabolite of rivastigmine and acts as an acetylcholinesterase (AChE) inhibitor. Supplied as a white to off-white solid reference standard, it is intended for biochemical research, Alzheimer's disease studies, and analytical applications requiring a characterized metabolite standard.
  • High purity (99.01%) suitable for analytical and reference use.
  • Well-defined chemical identity: C10H15NO, molecular weight 165.23 g·mol⁻1.
  • Solid form with clear appearance for straightforward handling and storage.
  • Recommended storage conditions provided for long-term stability.
  • Available in multiple package sizes to support both small-scale and larger studies.
Catalog No. 50-003-03356 Supplier Medchemexpress LLC Supplier No. 25GHYW020576
Rivastigmine metabolite (NAP 226-90), 3-[(1S)-1-(dimethylamino)ethyl]phenol, is the primary active metabolite of rivastigmine and acts as an acetylcholinesterase (AChE) inhibitor. Supplied as a white to off-white solid reference standard, it is intended for biochemical research, Alzheimer's disease studies, and analytical applications requiring a characterized metabolite standard.
  • High purity (99.01%) suitable for analytical and reference use.
  • Well-defined chemical identity: C10H15NO, molecular weight 165.23 g·mol⁻1.
  • Solid form with clear appearance for straightforward handling and storage.
  • Recommended storage conditions provided for long-term stability.
  • Available in multiple package sizes to support both small-scale and larger studies.