Learn More
Medchemexpress LLC 3-[(1S)-1-(dimethylamino)ethyl]phenol | 139306-10-8 | MFCD06656491 | 99.0% | 165.23 g/mol | C10H15NO | 250 MG
SDP
Catalog No. 5000302390
Encompass_Preferred
Rivastigmine metabolite (NAP 226-90) is the primary phenolic metabolite of rivastigmine and functions as an acetylcholinesterase (AChE) inhibitor. It is used as a reference standard and research reagent in studies of rivastigmine metabolism and Alzheimer's disease, supplied as a solid with high reported purity and available in multiple pack sizes including small assay-ready DMSO solutions.
  • High purity suitable for analytical standards and reference use.
  • Acts as an acetylcholinesterase inhibitor for biochemical studies.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes, including 250 mg and larger quantities.
  • Offered in DMSO solution formats for assay-ready applications.
Catalog No. 50-003-02390 Supplier Medchemexpress LLC Supplier No. 250MGHYW020576
Rivastigmine metabolite (NAP 226-90) is the primary phenolic metabolite of rivastigmine and functions as an acetylcholinesterase (AChE) inhibitor. It is used as a reference standard and research reagent in studies of rivastigmine metabolism and Alzheimer's disease, supplied as a solid with high reported purity and available in multiple pack sizes including small assay-ready DMSO solutions.
  • High purity suitable for analytical standards and reference use.
  • Acts as an acetylcholinesterase inhibitor for biochemical studies.
  • Solid, white to off-white appearance for easy handling.
  • Available in multiple pack sizes, including 250 mg and larger quantities.
  • Offered in DMSO solution formats for assay-ready applications.