Learn More
Medchemexpress LLC 3-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)propanoic acid | 784105-33-5 | MFCD21607085 | 99.9% | 221.25 g/mol | C9H19NO5 | 5 G
SDP
Catalog No. 5000204296
Encompass_Preferred
Amino-PEG3-C2-acid is a PEG-based bifunctional linker bearing a terminal amino group and a C2 carboxylic acid for use in antibody-drug conjugate and PROTAC synthesis and general bioconjugation.
  • Provides terminal amino and carboxylic acid groups for facile conjugation.
  • PEG3 spacer improves solubility and molecular flexibility.
  • High purity material suitable for synthetic and analytical workflows.
  • Available in multiple pack sizes, including 5 G, for small-scale and preparative use.
  • Storage recommendations: store neat at 4°C; in solvent store at -20°C (short term) or -80°C (long term).
Catalog No. 50-002-04296 Supplier Medchemexpress LLC Supplier No. HYW0401655G
Amino-PEG3-C2-acid is a PEG-based bifunctional linker bearing a terminal amino group and a C2 carboxylic acid for use in antibody-drug conjugate and PROTAC synthesis and general bioconjugation.
  • Provides terminal amino and carboxylic acid groups for facile conjugation.
  • PEG3 spacer improves solubility and molecular flexibility.
  • High purity material suitable for synthetic and analytical workflows.
  • Available in multiple pack sizes, including 5 G, for small-scale and preparative use.
  • Storage recommendations: store neat at 4°C; in solvent store at -20°C (short term) or -80°C (long term).