Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(2,2,2-Trifluoroacetamido)propanoic acid

Catalog No. OTV000473 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000473 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000473 Supplier Sigma Aldrich Supplier No. OTV000473250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triisopropyl phosphate, an ester of secondary alcohol, is a non-phosphorylating organophosphorus compound
  • It is formed as a by-product during the synthesis of haloalkylphosphonates
  • Triisopropyl phosphate can be prepared from isopropyl alcohol by reacting with phosphorus oxychloride
  • It can also be obtained from triisopropyl phosphite

Specifications

CAS 50632-82-1
Molecular Formula C5H6F3NO3
Linear Formula C5H6F3NO3
MDL Number MFCD00460921
Quantity 100 g
Molecular Weight (g/mol) 185.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.