Learn More
Sigma Organic Chemistry 3-2-2 4-Dinitrophenyl a | 100mg | MFCD28096676
Supplier: Sigma Organic Chemistry ANV00317100MG

3-2-2 4-Dinitrophenyl a , 100mg
About this Item:
MDL #: MFCD28096676
Molecular Weight: 299.24
UNSPSC Code: 12352200
Chemical Formula: C11H13N3O7
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.