Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane, tech.

Catalog No. AC346170250
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC346170250 25 mL Glass bottle
AC346171000 100 mL Glass bottle
AC346175000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC346170250 Supplier Thermo Scientific Chemicals Supplier No. 346170250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 35141-30-1 | C10H27N3O3Si | 265.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35141-30-1
Molecular Formula C10H27N3O3Si
Molecular Weight (g/mol) 265.43
MDL Number MFCD00069173
InChI Key NHBRUUFBSBSTHM-UHFFFAOYSA-N
Synonym n1-2-aminoethyl-n2-3-trimethoxysilyl propyl ethane-1,2-diamine, 3-trimethoxysilylpropyl diethylenetriamine, 3-2-2-aminoethylamino ethylamino propyl-trimethoxysilane, 3-trimethoxysilyl propyl diethylenetriamine, n1-3-trimethoxysilylpropyl diethylenetriamine, trimethoxysilylpropyldiethylenetriamine, 1,2-ethanediamine, n-2-aminoethyl-n'-3-trimethoxysilyl propyl, diethylenetriamino propyltrimethoxysilane, 3-2-2-aminoethylamino ethylamino propyltrimethoxysilane, n-2-aminoethyl-n'-3-trimethoxysilyl propyl ethylenediamine
PubChem CID 118771
IUPAC Name N'-[2-(3-trimethoxysilylpropylamino)ethyl]ethane-1,2-diamine
SMILES CO[Si](CCCNCCNCCN)(OC)OC

Specifications

Color Yellow
Density 1.0310g/mL
Boiling Point 114.0°C to 118.0°C (2.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4570 to 1.4610
Linear Formula (H3CO)3Si(CH2)3NH(CH2)2NH(CH2)2NH2
Quantity 25 mL
Specific Gravity 1.031
Solubility Information Solubility in water: immiscible
Formula Weight 265.43
Percent Purity 14 to 18% (amine (as NH2))
Grade Technical
Physical Form Liquid
Chemical Name or Material 3-[2-(2-Aminoethylamino)ethylamino]propyl-trimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
EINECSNumber 252-390-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.