Learn More
Medchemexpress LLC 3-[2,2-bis[(9Z,12Z)-octadeca-9,12-dienyl]-1,3-dioxolan-4-yl]-N,N-dimethylpropan-1-amine | 1217306-46-1 | >98.0% | C44H81NO2 | 10MG
SDP
Catalog No. 5000261464
Encompass_Preferred
A synthetic cationic lipid used in the formulation of nucleic acid-lipid particles (LNPs) for research delivery of RNA and DNA. It promotes encapsulation and cellular uptake of nucleic acids when combined with helper lipids and is supplied for laboratory research use only.
  • Cationic lipid for formulation of nucleic acid-lipid particles.
  • Facilitates encapsulation and delivery of RNA and DNA.
  • High purity suitable for research applications.
  • Powder form, soluble in DMSO for preparation of stock solutions.
  • Recommended storage at -20°C to maintain stability.
Catalog No. 50-002-61464 Supplier Medchemexpress LLC Supplier No. HY14522510MG
A synthetic cationic lipid used in the formulation of nucleic acid-lipid particles (LNPs) for research delivery of RNA and DNA. It promotes encapsulation and cellular uptake of nucleic acids when combined with helper lipids and is supplied for laboratory research use only.
  • Cationic lipid for formulation of nucleic acid-lipid particles.
  • Facilitates encapsulation and delivery of RNA and DNA.
  • High purity suitable for research applications.
  • Powder form, soluble in DMSO for preparation of stock solutions.
  • Recommended storage at -20°C to maintain stability.