Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[2-(2-methoxyethoxy)ethoxy]propanal | 356066-46-1 | MFCD26910955 | 99.8% | 176.21 g/mol | C8H16O4 | 10MG
SDP

Catalog No. 5000238898
Encompass_Preferred

m-PEG3-aldehyde is a short polyethylene glycol (PEG)-based linker bearing a methoxy terminus and a reactive aldehyde group. It is supplied for research use and is used to introduce a flexible PEG3 spacer and an aldehyde functional handle in the synthesis of PROTACs, bioconjugates, and other medicinal chemistry applications. The compound is typically handled and stored under inert conditions and is soluble in DMSO.

  • Provides a PEG3 spacer for flexible linkage.
  • Contains a terminal aldehyde for chemoselective conjugation to hydrazide or aminooxy groups.
  • Suitable for PROTAC synthesis and bioconjugation workflows.
  • High reported purity for research applications.
  • Soluble in DMSO for preparation of stock solutions.

Catalog No. 50-002-38898 Supplier Medchemexpress LLC Supplier No. HY12401110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG3-aldehyde is a short polyethylene glycol (PEG)-based linker bearing a methoxy terminus and a reactive aldehyde group. It is supplied for research use and is used to introduce a flexible PEG3 spacer and an aldehyde functional handle in the synthesis of PROTACs, bioconjugates, and other medicinal chemistry applications. The compound is typically handled and stored under inert conditions and is soluble in DMSO.

  • Provides a PEG3 spacer for flexible linkage.
  • Contains a terminal aldehyde for chemoselective conjugation to hydrazide or aminooxy groups.
  • Suitable for PROTAC synthesis and bioconjugation workflows.
  • High reported purity for research applications.
  • Soluble in DMSO for preparation of stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.