Learn More
Medchemexpress LLC 3-[2-(2-methoxyethoxy)ethoxy]propanal | 356066-46-1 | MFCD26910955 | 98.0% | 176.21 g/mol | C8H16O4 | 100MG
SDP
Catalog No. 5000235036
Encompass_Preferred
m-PEG3-aldehyde (stabilized with BHT) is a short polyethylene glycol (PEG) linker bearing a terminal aldehyde for chemoselective conjugation. Stabilized with butylated hydroxytoluene (BHT) to improve shelf stability, it is provided as a high-purity research reagent for use in PROTAC synthesis, bioconjugation, and related organic syntheses. Typical supply formats support small-scale and preparative workflows.
  • Terminal aldehyde functional group for chemoselective conjugation
  • Short PEG spacer provides flexibility and solubility
  • Stabilized with BHT to reduce oxidative degradation
  • High purity suitable for research and synthesis
  • Available in multiple pack sizes for scale flexibility
Catalog No. 50-002-35036 Supplier Medchemexpress LLC Supplier No. HY124011A100MG
m-PEG3-aldehyde (stabilized with BHT) is a short polyethylene glycol (PEG) linker bearing a terminal aldehyde for chemoselective conjugation. Stabilized with butylated hydroxytoluene (BHT) to improve shelf stability, it is provided as a high-purity research reagent for use in PROTAC synthesis, bioconjugation, and related organic syntheses. Typical supply formats support small-scale and preparative workflows.
  • Terminal aldehyde functional group for chemoselective conjugation
  • Short PEG spacer provides flexibility and solubility
  • Stabilized with BHT to reduce oxidative degradation
  • High purity suitable for research and synthesis
  • Available in multiple pack sizes for scale flexibility