Learn More
Medchemexpress LLC 3-[2-(2-methoxyethoxy)ethoxy]propanal | 356066-46-1 | MFCD26910955 | 99.8% | 176.21 g/mol | C8H16O4 | 5MG
SDP
Catalog No. 5000236284
Encompass_Preferred
m-PEG3-aldehyde is a methoxy-terminated polyethylene glycol linker bearing a terminal aldehyde functional group. It provides a short PEG spacer (three ethylene glycol units) to improve solubility and flexibility and enables conjugation by oxime formation or reductive amination for medicinal chemistry applications such as PROTAC assembly and bioconjugation.
  • Provides a reactive aldehyde for oxime formation and reductive amination.
  • Contains a short PEG spacer of three ethylene glycol units.
  • Improves solubility and conformational flexibility of conjugates.
  • Suitable for PROTAC linker synthesis and other bioconjugation workflows.
  • Supplied at research scale for laboratory synthesis.
  • High analytical purity (>99.8%) as reported by the manufacturer.
Catalog No. 50-002-36284 Supplier Medchemexpress LLC Supplier No. HY1240115MG
m-PEG3-aldehyde is a methoxy-terminated polyethylene glycol linker bearing a terminal aldehyde functional group. It provides a short PEG spacer (three ethylene glycol units) to improve solubility and flexibility and enables conjugation by oxime formation or reductive amination for medicinal chemistry applications such as PROTAC assembly and bioconjugation.
  • Provides a reactive aldehyde for oxime formation and reductive amination.
  • Contains a short PEG spacer of three ethylene glycol units.
  • Improves solubility and conformational flexibility of conjugates.
  • Suitable for PROTAC linker synthesis and other bioconjugation workflows.
  • Supplied at research scale for laboratory synthesis.
  • High analytical purity (>99.8%) as reported by the manufacturer.