Learn More
Medchemexpress LLC 3-(2-(2-methoxyethoxy)ethoxy)propanal | 356066-46-1 | MFCD26910955 | 99.8% | 176.21 g/mol | C8H16O4 | 50MG
SDP
Catalog No. 5000238061
Encompass_Preferred
m-PEG3-aldehyde is a PEG-based PROTAC linker consisting of a methoxy-terminated triethylene glycol chain with a terminal aldehyde functional group suitable for aldehyde-specific coupling chemistries, including reductive amination. It is provided as a small-scale research reagent for use in linker construction and bioconjugation applications.
  • High purity (99.8%).
  • Molecular weight 176.21 g/mol, formula C8H16O4.
  • Terminal aldehyde enables reductive amination and related conjugations.
  • PEG3 spacer improves solubility and flexibility in bioconjugates.
  • Suitable for use in PROTAC synthesis and other linker applications.
  • Store under nitrogen at -20°C; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-38061 Supplier Medchemexpress LLC Supplier No. HY12401150MG
m-PEG3-aldehyde is a PEG-based PROTAC linker consisting of a methoxy-terminated triethylene glycol chain with a terminal aldehyde functional group suitable for aldehyde-specific coupling chemistries, including reductive amination. It is provided as a small-scale research reagent for use in linker construction and bioconjugation applications.
  • High purity (99.8%).
  • Molecular weight 176.21 g/mol, formula C8H16O4.
  • Terminal aldehyde enables reductive amination and related conjugations.
  • PEG3 spacer improves solubility and flexibility in bioconjugates.
  • Suitable for use in PROTAC synthesis and other linker applications.
  • Store under nitrogen at -20°C; in solvent store at -80°C for long-term stability.