Learn More
Medchemexpress LLC 3-[2-(2-methylpyridin-4-yl)-4,6-dihydropyrrolo[3,4-c]pyrazole-5-carbonyl]-4-propan-2-yloxybenzonitrile | 1820934-93-7 | 98.6% | 387.43 g/mol | C22H21N5O2 | 5 MG
SDP
Catalog No. 5000264083
Encompass_Preferred
GlyT1 Inhibitor 1 is a potent, selective inhibitor of the glycine transporter 1 (GlyT1) that inhibits rat GlyT1 with an IC50 of 38 nM. It is supplied as a research reagent with reported antipsychotic activity, defined chemical identity, and high purity.
  • Selective GlyT1 inhibition (IC50 = 38 nM for rGlyT1).
  • Reported antipsychotic activity in preclinical studies.
  • High purity (98.6%) suitable for research applications.
  • Molecular formula C22H21N5O2; molecular weight 387.43 g/mol.
  • Available in multiple pack sizes and provided as DMSO solution or solid.
  • Intended for research use only, not for human or veterinary use.
Catalog No. 50-002-64083 Supplier Medchemexpress LLC Supplier No. HY1124325MG
GlyT1 Inhibitor 1 is a potent, selective inhibitor of the glycine transporter 1 (GlyT1) that inhibits rat GlyT1 with an IC50 of 38 nM. It is supplied as a research reagent with reported antipsychotic activity, defined chemical identity, and high purity.
  • Selective GlyT1 inhibition (IC50 = 38 nM for rGlyT1).
  • Reported antipsychotic activity in preclinical studies.
  • High purity (98.6%) suitable for research applications.
  • Molecular formula C22H21N5O2; molecular weight 387.43 g/mol.
  • Available in multiple pack sizes and provided as DMSO solution or solid.
  • Intended for research use only, not for human or veterinary use.