Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[2-(2-{[(tert-butoxy)carbonyl]amino}ethoxy)ethoxy]propanoic acid | 1365655-91-9 | MFCD22056304 | 98.0% | 277.31 | C12H23NO6 | 5 G
SDP

Catalog No. 5000300125
Encompass_Preferred

Boc-NH-PEG2-CH2CH2COOH is a Boc-protected PEG2 linker containing a terminal carboxylic acid and a protected amine. It is designed as a short, two-unit PEG spacer for use in the synthesis of PROTACs and bifunctional conjugates, supplied for research use with documented solubility and storage recommendations.

  • Boc-protected amino group for orthogonal deprotection
  • Two-unit PEG spacer to improve solubility and linker flexibility
  • Terminal carboxylic acid for amide or ester coupling reactions
  • Molecular weight 277.31 and formula C12H23NO6
  • High solubility in DMSO (≥100 mg/mL) and formulatable in several vehicles
  • Storage stability: long-term at -20°C; shorter stability in solution
  • For research use only; not validated for medical applications

Catalog No. 50-003-00125 Supplier Medchemexpress LLC Supplier No. HYW0258125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG2-CH2CH2COOH is a Boc-protected PEG2 linker containing a terminal carboxylic acid and a protected amine. It is designed as a short, two-unit PEG spacer for use in the synthesis of PROTACs and bifunctional conjugates, supplied for research use with documented solubility and storage recommendations.

  • Boc-protected amino group for orthogonal deprotection
  • Two-unit PEG spacer to improve solubility and linker flexibility
  • Terminal carboxylic acid for amide or ester coupling reactions
  • Molecular weight 277.31 and formula C12H23NO6
  • High solubility in DMSO (≥100 mg/mL) and formulatable in several vehicles
  • Storage stability: long-term at -20°C; shorter stability in solution
  • For research use only; not validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.