Learn More
Medchemexpress LLC 3-(2-(4-(2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-5-yl)piperazin-1-yl)ethoxy)propanoic acid | 2731007-11-5 | 99.2% | C22H26N4O7 | 10MG
SDP
Catalog No. 5000262030
Encompass_Preferred
Thalidomide-Piperazine-PEG1-COOH is an E3 ligase ligand-linker conjugate combining a thalidomide-derived cereblon ligand with a short PEG1-piperazine linker terminated by a carboxylic acid. It is provided as a solid research reagent intended for use as a CRBN-binding module in PROTAC design and conjugation studies.
  • Enables CRBN engagement via a thalidomide-derived ligand.
  • Features a PEG1-piperazine linker with a terminal carboxylic acid for conjugation.
  • High reported purity enables reliable analytical and synthetic work.
  • Solid, white-to-yellow appearance for easy handling and storage.
  • Recommended storage at -20°C under inert gas to preserve integrity.
  • Available in small milligram quantities suitable for early-stage research.
Catalog No. 50-002-62030 Supplier Medchemexpress LLC Supplier No. HY13878210MG
Thalidomide-Piperazine-PEG1-COOH is an E3 ligase ligand-linker conjugate combining a thalidomide-derived cereblon ligand with a short PEG1-piperazine linker terminated by a carboxylic acid. It is provided as a solid research reagent intended for use as a CRBN-binding module in PROTAC design and conjugation studies.
  • Enables CRBN engagement via a thalidomide-derived ligand.
  • Features a PEG1-piperazine linker with a terminal carboxylic acid for conjugation.
  • High reported purity enables reliable analytical and synthetic work.
  • Solid, white-to-yellow appearance for easy handling and storage.
  • Recommended storage at -20°C under inert gas to preserve integrity.
  • Available in small milligram quantities suitable for early-stage research.