Learn More
Medchemexpress LLC 3-[[2-[[(4-Carbamimidoylphenyl)amino]methyl]-1-(trideuteriomethyl)benzimidazole-5-carbonyl]-pyridin- | 1246817-44-6 | MFCD21363573 | 95.1% | 474.53 | C25H22D3N7O3 | 10 MG
SDP
Catalog No. 5000245661
Encompass_Preferred
Dabigatran-d3 is the deuterated (d3) stable-isotope analog of the direct thrombin inhibitor dabigatran. Supplied as an off-white to light-yellow solid, it is intended as an analytical reference standard or internal standard for quantitative mass-spectrometry assays. CAS 1246817-44-6; formula C25H22D3N7O3; MW 474.53; HPLC purity 95.14%.
  • Deuterated internal standard for LC-MS quantitation.
  • Off-white to light-yellow solid suitable for storage and handling.
  • High isotopic labeling for accurate analyte normalization.
  • Characterized molecular weight and formula for method development.
  • Sufficient HPLC purity for analytical reference use.
Catalog No. 50-002-45661 Supplier Medchemexpress LLC Supplier No. HY10163S110MG
Dabigatran-d3 is the deuterated (d3) stable-isotope analog of the direct thrombin inhibitor dabigatran. Supplied as an off-white to light-yellow solid, it is intended as an analytical reference standard or internal standard for quantitative mass-spectrometry assays. CAS 1246817-44-6; formula C25H22D3N7O3; MW 474.53; HPLC purity 95.14%.
  • Deuterated internal standard for LC-MS quantitation.
  • Off-white to light-yellow solid suitable for storage and handling.
  • High isotopic labeling for accurate analyte normalization.
  • Characterized molecular weight and formula for method development.
  • Sufficient HPLC purity for analytical reference use.