Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[[[2,4-dicyano-3-[4-[2-(methylamino)-2-oxoethoxy]phenyl]pyrido[1,2-a]benzimidazol-1-yl]amino]carbonyl]-L-phenylalanine | 1941174-13-5 | 99.8% | 1 MG
SDP

Catalog No. 5000233941
Encompass_Preferred

KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter 1 (LAT1/SLC7A5). It inhibits uptake of LAT1 substrates such as L-leucine (reported IC50 ≈ 18 μM) and reduces proliferation in cancer cell models. Supplied as a high-purity research compound for in vitro studies; store powder at -20°C.

  • Potent, reversible, selective LAT1 inhibition (IC50 ≈ 18 μM).
  • High purity suitable for research (reported 99.75%).
  • Molecular formula C32H25N7O5; molecular weight ~587.6.
  • Storage: powder at -20°C; solutions -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for screening and assays (for example, 1 mg).

Catalog No. 50-002-33941 Supplier Medchemexpress LLC Supplier No. HY1201391MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter 1 (LAT1/SLC7A5). It inhibits uptake of LAT1 substrates such as L-leucine (reported IC50 ≈ 18 μM) and reduces proliferation in cancer cell models. Supplied as a high-purity research compound for in vitro studies; store powder at -20°C.

  • Potent, reversible, selective LAT1 inhibition (IC50 ≈ 18 μM).
  • High purity suitable for research (reported 99.75%).
  • Molecular formula C32H25N7O5; molecular weight ~587.6.
  • Storage: powder at -20°C; solutions -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for screening and assays (for example, 1 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.