Learn More
Medchemexpress LLC 3-[2,4-dimethyl-5-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid | 210644-62-5 | MFCD09743433 | 99.4% | 310.35 | C18H18N2O3 | 50 MG
SDP
Catalog No. 5000242440
Encompass_Preferred
Orantinib is a Z-isomer small-molecule tyrosine kinase inhibitor that targets VEGFR (Flk-1/KDR), PDGFRβ, and FGFR1. Supplied as an orange-red powder for research use, it is used in preclinical studies of angiogenesis and receptor tyrosine kinase signaling. Molecular formula C18H18N2O3; molecular weight 310.35; CAS 210644-62-5.
  • Inhibits VEGFR, PDGFRβ, and FGFR1 for angiogenesis research.
  • Orally active small-molecule inhibitor suitable for cell and biochemical assays.
  • High reported purity supports reproducible experimental results.
  • Solid powder form with defined storage conditions for long-term stability.
  • Available in multiple pack sizes to support varied study scales.
Catalog No. 50-002-42440 Supplier Medchemexpress LLC Supplier No. HY10517A50MG
Orantinib is a Z-isomer small-molecule tyrosine kinase inhibitor that targets VEGFR (Flk-1/KDR), PDGFRβ, and FGFR1. Supplied as an orange-red powder for research use, it is used in preclinical studies of angiogenesis and receptor tyrosine kinase signaling. Molecular formula C18H18N2O3; molecular weight 310.35; CAS 210644-62-5.
  • Inhibits VEGFR, PDGFRβ, and FGFR1 for angiogenesis research.
  • Orally active small-molecule inhibitor suitable for cell and biochemical assays.
  • High reported purity supports reproducible experimental results.
  • Solid powder form with defined storage conditions for long-term stability.
  • Available in multiple pack sizes to support varied study scales.