Learn More
Medchemexpress LLC 3-[2,4-dimethyl-5-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid | 252916-29-3 | MFCD09743433 | >98.0% | 310.3 g/mol | C18H18N2O3 | 200 MG
SDP
Catalog No. 5000243943
Encompass_Preferred
Orantinib is a small-molecule, orally bioavailable receptor tyrosine kinase inhibitor that selectively inhibits VEGFR2, PDGFRβ, and FGFR1, blocking receptor autophosphorylation and angiogenesis. It is used in preclinical and clinical research for tumor growth inhibition and antiangiogenic studies. Chemical formula C18H18N2O3; molecular weight ~310.3 g/mol; CAS 252916-29-3.
  • Multi-target inhibition of VEGFR2, PDGFRβ, and FGFR1.
  • Orally bioavailable small molecule suitable for in vivo and in vitro studies.
  • Blocks receptor autophosphorylation and inhibits angiogenesis.
  • Applied in oncology and angiogenesis research applications.
  • High purity material appropriate for research use.
Catalog No. 50-002-43943 Supplier Medchemexpress LLC Supplier No. HY10517200MG
Orantinib is a small-molecule, orally bioavailable receptor tyrosine kinase inhibitor that selectively inhibits VEGFR2, PDGFRβ, and FGFR1, blocking receptor autophosphorylation and angiogenesis. It is used in preclinical and clinical research for tumor growth inhibition and antiangiogenic studies. Chemical formula C18H18N2O3; molecular weight ~310.3 g/mol; CAS 252916-29-3.
  • Multi-target inhibition of VEGFR2, PDGFRβ, and FGFR1.
  • Orally bioavailable small molecule suitable for in vivo and in vitro studies.
  • Blocks receptor autophosphorylation and inhibits angiogenesis.
  • Applied in oncology and angiogenesis research applications.
  • High purity material appropriate for research use.