Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[2-(4-hydroxyphenyl)ethyl]phenol | 37116-80-6 | MFCD24713452 | 97.0% | 214.26 g/mol | C14H14O2 | 250MG
SDP

Catalog No. 5000238951
Encompass_Preferred

Lunularin is a small-molecule research chemical reported as an 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) inhibitor and an endogenous metabolite. It is supplied for laboratory and analytical use only. Identifiers: CAS 37116-80-6; formula C14H14O2; molecular weight 214.26 g/mol; listed purity 97.0%.

  • Small-molecule 11β-HSD1 inhibitor and endogenous metabolite.
  • Intended for research and analytical applications only.
  • Purity 97.0% as supplied.
  • Molecular weight 214.26 g/mol; formula C14H14O2.
  • Available in multiple pack sizes, including a 250 mg pack.

Catalog No. 50-002-38951 Supplier Medchemexpress LLC Supplier No. HY124529250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lunularin is a small-molecule research chemical reported as an 11β-hydroxysteroid dehydrogenase 1 (11β-HSD1) inhibitor and an endogenous metabolite. It is supplied for laboratory and analytical use only. Identifiers: CAS 37116-80-6; formula C14H14O2; molecular weight 214.26 g/mol; listed purity 97.0%.

  • Small-molecule 11β-HSD1 inhibitor and endogenous metabolite.
  • Intended for research and analytical applications only.
  • Purity 97.0% as supplied.
  • Molecular weight 214.26 g/mol; formula C14H14O2.
  • Available in multiple pack sizes, including a 250 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.