Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-(2,5,6-Trimethyl-4-oxothieno[2,3-d]pyrimidin-3(4H)-yl)propionic acid

Catalog No. OTV000559 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV000559 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV000559 Supplier Sigma Aldrich Supplier No. OTV000559250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-trifluoromethyl-2-propanol is a fluorinated aliphatic alcohol
  • It can be prepared by reacting methyllithium, trifluoroacetone and trifluorovinyl bromide in ether
  • This can also be synthesized from the reaction between methylmagnesium bromide and 1,1,1-trifluoroacetone in ether.2-trifluoromethyl-2-propanol may be employed as a solvent for the preparation of [18f]fluorothymidine
  • It may also be used in the synthesis of [2-(trifluoromethyl)-2-propyl nitrate] through nitration in nitric acid/trifluoroacetic anhydride

Specifications

Molecular Formula C12H14N2O3S
Linear Formula C12H14N2O3S
MDL Number MFCD09905323
Quantity 5 g
Molecular Weight (g/mol) 266.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.