Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride, Thermo Scientific™

Catalog No. SB00576DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB00576DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB00576DA Supplier Thermo Scientific Chemicals Supplier No. SB00576DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4462-55-9
Molecular Formula C11H6Cl3NO2
Molecular Weight (g/mol) 290.524
InChI Key IZQGELJKDARDMZ-UHFFFAOYSA-N
Synonym 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonyl chloride, dcimc chloride, 3-2,6-dichlorophenyl-5-methyl-1,2-oxazole-4-carbonyl chloride, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carbonylchloride, 4-isoxazolecarbonyl chloride, 3-2,6-dichlorophenyl-5-methyl, 3-2,6-dichlorophenyl-5-methylisoxazole-4-carboxylic acid chloride, 3-2,6-dichlorophenyl-5-methyl isoxazole-4-carbonyl chloride, pubchem8683, dsstox_cid_31536, dsstox_rid_97421
PubChem CID 78212
IUPAC Name 3-(2,6-dichlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C2=C(C=CC=C2Cl)Cl)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 290.53
Percent Purity 97%
Chemical Name or Material 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.