Learn More
Medchemexpress LLC 3-[(2,7-dimethoxyacridin-9-yl)sulfanyl]propan-1-amine | 184582-62-5 | MFCD02752814 | 99.6% | C18H20N2O2S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1345510MG
Encompass_Preferred
LDN-192960 is a small-molecule research inhibitor selective for Haspin kinase and Dual-specificity Tyrosine-regulated Kinase 2 (DYRK2), exhibiting low-nanomolar potency in enzymatic assays. It is provided for laboratory research as a high-purity solid or as a 10 mM solution in DMSO and is used in studies of cell-cycle regulation and kinase signaling.
  • Potent inhibition of Haspin (IC50 ≈ 10 nM) and DYRK2 (IC50 ≈ 48 nM).
  • High reported purity (≈99.6%).
  • Available as solid quantities and as a 10 mM DMSO solution for assay use.
  • Molecular formula C18H20N2O2S; molecular weight ≈328.43 g/mol.
  • Intended for laboratory research use only; not for human or clinical use.
Catalog No. 50-002-61758