Learn More
eMolecules​ [3-(2-Adamantyl)-4-methoxyphenyl]magnesium bromide, 0.5 M in THF | Synthonix |345.563 | C17H21BrMgO | 0.000 | [Br-].COc1ccc([Mg+])cc1C1C2CC3CC(C2)CC1C3 | 50ml | 818571991
Catalog No. 502694376 Shop All eMolecules Inc. Products
Encompass_Preferred
[3-(2-Adamantyl)-4-methoxyphenyl]magnesium bromide, 0.5 M in THF | Synthonix |345.563 | C17H21BrMgO | 0.000 | [Br-].COc1ccc([Mg+])cc1C1C2CC3CC(C2)CC1C3 | 50ml | 818571991
Catalog No. 50-269-4376 Supplier eMolecules​ Supplier No. 818571991
[3-(2-Adamantyl)-4-methoxyphenyl]magnesium bromide, 0.5 M in THF | Synthonix |345.563 | C17H21BrMgO | 0.000 | [Br-].COc1ccc([Mg+])cc1C1C2CC3CC(C2)CC1C3 | 50ml | 818571991