Learn More
Medchemexpress LLC 3-[(2-aminoethyl)dithio]propionic acid | 15579-00-7 | MFCD07357249 | 98.0% | 181.28 g/mol | C5H11NO2S2 | 25 MG
SDP
Catalog No. 5000202530
Encompass_Preferred
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the synthesis of antibody-drug conjugates (ADCs). It provides a reducible linkage that allows intracellular release of cytotoxic payloads under reducing conditions; the compound is supplied as a solid with analytical documentation (COA, SDS, HNMR, LCMS).
  • Cleavable disulfide linker enabling intracellular payload release.
  • High purity suitable for research use (≈98% by HPLC).
  • Multiple pack sizes available from milligram to gram scale.
  • Characterized by HNMR and LCMS, with COA and SDS available.
  • Functional groups: primary amine, carboxylic acid, and disulfide for bioconjugation.
Catalog No. 50-002-02530 Supplier Medchemexpress LLC Supplier No. HY14009625MG
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used in the synthesis of antibody-drug conjugates (ADCs). It provides a reducible linkage that allows intracellular release of cytotoxic payloads under reducing conditions; the compound is supplied as a solid with analytical documentation (COA, SDS, HNMR, LCMS).
  • Cleavable disulfide linker enabling intracellular payload release.
  • High purity suitable for research use (≈98% by HPLC).
  • Multiple pack sizes available from milligram to gram scale.
  • Characterized by HNMR and LCMS, with COA and SDS available.
  • Functional groups: primary amine, carboxylic acid, and disulfide for bioconjugation.