Learn More
Medchemexpress LLC 3-(2-Aminoethyl)indole | 61-54-1 | 25 MG
SDP
Catalog No. 5000413569
Encompass_Preferred
Tryptamine is a selective, blood-brain-penetrating 5-HT4 receptor agonist and an endogenous ligand of the aryl hydrocarbon receptor (AHR). It promotes intestinal anion secretion and fluid transport, accelerating gastrointestinal motility. Tryptamine also regulates Th17/Treg balance to inhibit neuroinflammation and participates in temperature regulation as a neuromodulator. It is used to study intestinal disorders and has shown efficacy in multiple sclerosis models.
  • Selective 5-HT4 receptor agonist and AHR ligand
  • Promotes intestinal anion secretion and fluid transport
  • Accelerates gastrointestinal motility
  • Regulates Th17/Treg balance to inhibit neuroinflammation
  • Participates in temperature regulation and spinal reflex regulation
  • Used to study intestinal disorders
  • Shown efficacy in multiple sclerosis models
Catalog No. 50-004-13569 Supplier Medchemexpress LLC Supplier No. 25MGHYB2132R
Tryptamine is a selective, blood-brain-penetrating 5-HT4 receptor agonist and an endogenous ligand of the aryl hydrocarbon receptor (AHR). It promotes intestinal anion secretion and fluid transport, accelerating gastrointestinal motility. Tryptamine also regulates Th17/Treg balance to inhibit neuroinflammation and participates in temperature regulation as a neuromodulator. It is used to study intestinal disorders and has shown efficacy in multiple sclerosis models.
  • Selective 5-HT4 receptor agonist and AHR ligand
  • Promotes intestinal anion secretion and fluid transport
  • Accelerates gastrointestinal motility
  • Regulates Th17/Treg balance to inhibit neuroinflammation
  • Participates in temperature regulation and spinal reflex regulation
  • Used to study intestinal disorders
  • Shown efficacy in multiple sclerosis models