Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [3-(2-Aminoethylamino)Propyl]Trimethoxysilane

Catalog No. 440302100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-030-2100ML 100 mL
44-030-2500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 44-030-2100ML Supplier Sigma Aldrich Supplier No. 440302100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[3-(2-Aminoethylamino)propyl]trimethoxysilane has been used in the synthesis of various polyhedral silsesquioxanes, (RSiO1.5)n (SSQO).It can be functionalized onto the mesoporous SBA-15 silica to be used as matrices for controlled drug delivery.It is also used in the synthesis of silane-based polymer, which is used in conducting instant adhesives.Repackaged by Aldrich Chemical Company, Inc. Manufactured by Dow Corning Corporation, U.S.A.

Specifications

CAS 1760-24-3
Density 1.028 g/mL at 25°C
Boiling Point 146°C (15 mmHg, lit.)
Molecular Formula C8H22N2O3Si
Linear Formula (CH3O)3Si(CH2)3NHCH2CH2NH2
Refractive Index n20/D 1.444
Quantity 100 mL
MDL Number MFCD00008173
Synonym Dow Corning(R) product Z-6020; N-(2-Aminoethyl)-3-(trimethoxysilyl)propylamine
Molecular Weight (g/mol) 222.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.