Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Aminoethylamino)propylamine, 97%, Thermo Scientific™

Catalog No. AAA1910888
Change view
Click to view available options
Quantity:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1910888 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1910888 Supplier Thermo Scientific Chemicals Supplier No. A1910888

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13531-52-7
Flash Point 96°C (205°F)
Odor Amine-like
Molecular Formula C5H15N3
Quantity 2.5g
MDL Number MFCD00008211
UN Number UN2735
Synonym n-2-aminoethyl-1,3-propanediamine, 1,3-propanediamine, n-2-aminoethyl, unii-4eow03ix8s, n-3-aminopropyl ethylenediamine, 2-aminoethyl 3-aminopropyl amine, 4eow03ix8s, 1,3-propanediamine, n1-2-aminoethyl, n-2-aminoethyl propane-1,3-diamine, n1-2-aminoethyl propane-1,3-diamine, 3-2-aminoethylamino propylamine
InChI Key DTSDBGVDESRKKD-UHFFFAOYSA-N
SMILES C(CN)CNCCN
IUPAC Name N'-(2-aminoethyl)propane-1,3-diamine
Molecular Weight (g/mol) 117.196
PubChem CID 26091
Formula Weight 117.2
Percent Purity 97%
Chemical Name or Material 3-(2-Aminoethylamino)propylamine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.