Learn More
Medchemexpress LLC 3-(2-aminoethyldisulfanyl)propanoic acid | 15579-00-7 | MFCD07357249 | 98.0% | 181.28 | C5H11NO2S2 | 1 G
SDP
Catalog No. 5000203817
Encompass_Preferred
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used as a reducible linker in the synthesis of antibody-drug conjugates (ADCs) for research applications. It has molecular formula C5H11NO2S2, molecular weight 181.28, and reported purity of 98.03%.
  • Cleavable disulfide linker suitable for intracellular release
  • Used for synthesis of antibody-drug conjugates and other reducible conjugates
  • Offered in multiple sizes, including 1 G, for research flexibility
  • High reported purity (98.03%) suitable for research use
  • Solid reagent with formula C5H11NO2S2 and molecular weight 181.28
Catalog No. 50-002-03817 Supplier Medchemexpress LLC Supplier No. HY1400961G
Aminoethyl-SS-propionic acid is a cleavable disulfide-containing linker used as a reducible linker in the synthesis of antibody-drug conjugates (ADCs) for research applications. It has molecular formula C5H11NO2S2, molecular weight 181.28, and reported purity of 98.03%.
  • Cleavable disulfide linker suitable for intracellular release
  • Used for synthesis of antibody-drug conjugates and other reducible conjugates
  • Offered in multiple sizes, including 1 G, for research flexibility
  • High reported purity (98.03%) suitable for research use
  • Solid reagent with formula C5H11NO2S2 and molecular weight 181.28