Learn More
Abcam 3-(2-Benzothiazolyl)-7-(diethylamino)coumarin, Fluorescent dye, 5G
Catalog No. 502705247
Encompass_Preferred
MW 350.43 g/mol, Purity >98%. Fluorescent dye widely used for staining polymer particles, but also organelles or materials used in medicine.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5247 Supplier Abcam Supplier No. AB1453815G
MW 350.43 g/mol, Purity >98%. Fluorescent dye widely used for staining polymer particles, but also organelles or materials used in medicine.
The product is subject to the following: Abcam Restricted Use Statement