Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Bromoethylamino)propylamine dihydrobromide, 97%, Thermo Scientific™

Catalog No. AAH60309MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60309MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60309MD Supplier Thermo Scientific Chemicals Supplier No. H60309MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23545-42-8
Molecular Formula C5H15Br3N2
Molecular Weight (g/mol) 342.901
MDL Number MFCD00460336
InChI Key QJWQDMBGXNKPAS-UHFFFAOYSA-N
Synonym n-2-bromoethyl-1,3-propanediamine dihydrobromide, n1-2-bromoethyl propane-1,3-diamine dihydrobromide, 3-aminopropyl 2-bromoethyl amine dihydrobromide, n1-2-bromoethyl propane-1,3-diamine 2hbr, 1,3-propanediamine,n-2-bromoethyl dihydrobromide, 2-3-aminopropylamino ethyl bromide dihydrobromide, n-2-bromoethyl-1,3-propanediaminedihydrobromide, 3-aminopropyl 2-bromoethyl amine, bromide, bromide, 3-2-bromoethylamino propylamine dihydrobromide
PubChem CID 13354961
IUPAC Name N'-(2-bromoethyl)propane-1,3-diamine;dihydrobromide
SMILES C(CN)CNCCBr.Br.Br

Specifications

Linear Formula BrCH2CH2NHCH2CH2CH2NH2·2HBr
Quantity 250 mg
Formula Weight 342.9
Percent Purity 97%
Chemical Name or Material 3-(2-Bromoethylamino)propylamine dihydrobromide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.