Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Bromophenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B5152200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5152200MG 200 mg
B51521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5152200MG Supplier TCI America Supplier No. B5152200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 847264-30-6
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD18415514
InChI Key XEQMSSLGEYCVDC-UHFFFAOYSA-N
PubChem CID 17836216
IUPAC Name 3-(2-bromophenyl)pyridine
SMILES C1=CC=C(C(=C1)C2=CN=CC=C2)Br

Specifications

Boiling Point 191°C
Quantity 200 mg
Formula Weight 234.10
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(2-Bromophenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.