Learn More
Medchemexpress LLC 3-[(2-chloro-6-fluorophenyl)methylsulfanyl]-6-pyridin-2-ylpyridazine | 892415-28-0 | 98.9% | 331.8 g/mol | C16H11ClFN3S | 5 MG
SDP
Catalog No. 5000221450
Encompass_Preferred
EAAT2 activator 1 is a small-molecule activator of the excitatory amino acid transporter 2 (EAAT2). It increases EAAT2 protein levels in a dose-dependent manner and is supplied for laboratory research applications in solid form or as a DMSO solution.
  • Small-molecule activator of EAAT2 for research use.
  • Increases EAAT2 protein levels dose-dependently.
  • High purity (98.9%).
  • Available in small solid quantities (e.g., 5 mg) and as 10 mM solution in DMSO.
  • Solid appearance: light brown to brown powder.
  • Storage recommendations provided for powder and solvent solutions.
Catalog No. 50-002-21450 Supplier Medchemexpress LLC Supplier No. HY1396925MG
EAAT2 activator 1 is a small-molecule activator of the excitatory amino acid transporter 2 (EAAT2). It increases EAAT2 protein levels in a dose-dependent manner and is supplied for laboratory research applications in solid form or as a DMSO solution.
  • Small-molecule activator of EAAT2 for research use.
  • Increases EAAT2 protein levels dose-dependently.
  • High purity (98.9%).
  • Available in small solid quantities (e.g., 5 mg) and as 10 mM solution in DMSO.
  • Solid appearance: light brown to brown powder.
  • Storage recommendations provided for powder and solvent solutions.