Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[(2-chloro-6-fluorophenyl)methylsulfanyl]-6-pyridin-2-ylpyridazine | 892415-28-0 | 98.9% | C16H11ClFN3S | 10MG
SDP

Catalog No. 5000261809
Encompass_Preferred

EAAT2 activator 1 is a small-molecule research compound that increases protein levels of the excitatory amino acid transporter 2 (EAAT2), enhancing glutamate clearance in preclinical and in vitro studies. Supplied in solid form and as a 10 mM DMSO solution, it is intended for laboratory research use only.

  • Potent activator of EAAT2, used to upregulate glutamate transporter levels.
  • Available as solid and 10 mM DMSO solution formats for flexible handling.
  • High purity (98.9%) suitable for biochemical and cell-based assays.
  • Chemical formula C16H11ClFN3S; molecular weight 331.8 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; solutions at -80°C or -20°C for short term.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-61809 Supplier Medchemexpress LLC Supplier No. HY13969210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EAAT2 activator 1 is a small-molecule research compound that increases protein levels of the excitatory amino acid transporter 2 (EAAT2), enhancing glutamate clearance in preclinical and in vitro studies. Supplied in solid form and as a 10 mM DMSO solution, it is intended for laboratory research use only.

  • Potent activator of EAAT2, used to upregulate glutamate transporter levels.
  • Available as solid and 10 mM DMSO solution formats for flexible handling.
  • High purity (98.9%) suitable for biochemical and cell-based assays.
  • Chemical formula C16H11ClFN3S; molecular weight 331.8 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; solutions at -80°C or -20°C for short term.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.