Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(2-chloroacetyl)-1H-indole-5-carbonitrile | 115027-08-2 | MFCD13173395 | 218.64 g/mol | C11H7ClN2O | 100 MG
SDP

Catalog No. 5000210356
Encompass_Preferred

3-(2-chloroacetyl)-1H-indole-5-carbonitrile is a chemical intermediate used in the synthesis of pharmaceutical compounds. It has molecular formula C11H7ClN2O and molecular weight 218.64 g/mol.

  • Drug intermediate for synthesis of various active compounds.
  • Molecular formula: C11H7ClN2O.
  • Molecular weight: 218.64 g/mol.
  • CAS number: 115027-08-2.
  • Available in 100 mg pack (supplier SKU indicates 100 mg).

Catalog No. 50-002-10356 Supplier Medchemexpress LLC Supplier No. HYW048002100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-(2-chloroacetyl)-1H-indole-5-carbonitrile is a chemical intermediate used in the synthesis of pharmaceutical compounds. It has molecular formula C11H7ClN2O and molecular weight 218.64 g/mol.

  • Drug intermediate for synthesis of various active compounds.
  • Molecular formula: C11H7ClN2O.
  • Molecular weight: 218.64 g/mol.
  • CAS number: 115027-08-2.
  • Available in 100 mg pack (supplier SKU indicates 100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.