Learn More
Medchemexpress LLC 3-(2-chloroacetyl)-1H-indole-5-carbonitrile | 115027-08-2 | MFCD13173395 | 218.64 g/mol | C11H7ClN2O | 250 MG
SDP
Catalog No. 5000209191
Encompass_Preferred
3-(2-Chloroacetyl)-1H-indole-5-carbonitrile is a small-molecule drug intermediate supplied as a solid powder for use in organic synthesis and medicinal chemistry research. It is used as a building block to prepare substituted indole derivatives and other active compounds. The material is provided in laboratory-scale sizes and should be stored under cold conditions to maintain stability.
  • Solid powder for synthetic use.
  • Used as a drug intermediate and building block in medicinal chemistry.
  • Available in multiple laboratory-scale sizes, including 250 mg.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C is acceptable.
  • CAS number 115027-08-2; molecular weight 218.64 g/mol.
Catalog No. 50-002-09191 Supplier Medchemexpress LLC Supplier No. 250MGHYW048002
3-(2-Chloroacetyl)-1H-indole-5-carbonitrile is a small-molecule drug intermediate supplied as a solid powder for use in organic synthesis and medicinal chemistry research. It is used as a building block to prepare substituted indole derivatives and other active compounds. The material is provided in laboratory-scale sizes and should be stored under cold conditions to maintain stability.
  • Solid powder for synthetic use.
  • Used as a drug intermediate and building block in medicinal chemistry.
  • Available in multiple laboratory-scale sizes, including 250 mg.
  • Store powder at -20°C for long-term stability; short-term storage at 4°C is acceptable.
  • CAS number 115027-08-2; molecular weight 218.64 g/mol.