Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-(2-chloropyrimidin-4-yl)-1-methylindole | 1032452-86-0 | 99.5% | C13H10ClN3 | 25 G
SDP

Catalog No. 5000358572
Encompass_Preferred

This product is 3-(2-chloropyrimidin-4-yl)-1-methylindole, a high-purity chemical compound supplied as an off-white to light yellow solid. With a molecular formula of C13H10ClN3, it is suitable for various research applications.

  • High purity: 99.5% (HPLC)
  • Appearance: Off-white to light yellow solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-58572 Supplier Medchemexpress LLC Supplier No. 25GHY78866
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is 3-(2-chloropyrimidin-4-yl)-1-methylindole, a high-purity chemical compound supplied as an off-white to light yellow solid. With a molecular formula of C13H10ClN3, it is suitable for various research applications.

  • High purity: 99.5% (HPLC)
  • Appearance: Off-white to light yellow solid
  • Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
  • Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.