Learn More
Medchemexpress LLC 3-[2-cyano-3-(trifluoromethyl)phenoxy]phenyl-4,4,4-trifluoro-1-butanesulfonate | 406205-74-1 | 98.6% | 453.36 g·mol⁻1 | C18H13F6NO4S | 50 MG
SDP
Catalog No. 5000214954
Encompass_Preferred
BAY 59-3074 is a cannabinoid CB1/CB2 receptor partial agonist originally reported by Bayer. It has been characterized in vitro and in vivo for analgesic and antihyperalgesic effects and is used in research to probe cannabinoid receptor pharmacology.
  • Selective CB1/CB2 receptor partial agonist (human CB1 Ki = 48.3 nM; human CB2 Ki = 45.5 nM).
  • High purity: 98.6%.
  • Chemical formula C18H13F6NO4S; molecular weight 453.36 g·mol⁻1.
  • Solid, white to off-white physical form, suitable for laboratory handling.
  • Documented in peer-reviewed literature and product datasheets for research use.
Catalog No. 50-002-14954 Supplier Medchemexpress LLC Supplier No. HY10048850MG
BAY 59-3074 is a cannabinoid CB1/CB2 receptor partial agonist originally reported by Bayer. It has been characterized in vitro and in vivo for analgesic and antihyperalgesic effects and is used in research to probe cannabinoid receptor pharmacology.
  • Selective CB1/CB2 receptor partial agonist (human CB1 Ki = 48.3 nM; human CB2 Ki = 45.5 nM).
  • High purity: 98.6%.
  • Chemical formula C18H13F6NO4S; molecular weight 453.36 g·mol⁻1.
  • Solid, white to off-white physical form, suitable for laboratory handling.
  • Documented in peer-reviewed literature and product datasheets for research use.