Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[(2-Fluoro-4-iodophenyl)amino]isonicotinic acid | 885588-03-4 | C12H8FIN2O2 | 50 MG
SDP

Catalog No. 5000433818
Encompass_Preferred

3-[(2-Fluoro-4-iodophenyl)amino]isonicotinic acid is a chemical compound that serves as a drug intermediate, crucial for the synthesis of various active compounds. Its precise molecular structure and properties make it suitable for advanced chemical synthesis applications.

  • Functions as a drug intermediate
  • Essential for synthesizing various active compounds
  • Molecular weight of 358.11
  • Recommended storage at 4°C, protected from light
  • Solvent storage conditions include -80°C for six months or -20°C for one month, both protected from light

Catalog No. 50-004-33818 Supplier Medchemexpress LLC Supplier No. HY6601650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

3-[(2-Fluoro-4-iodophenyl)amino]isonicotinic acid is a chemical compound that serves as a drug intermediate, crucial for the synthesis of various active compounds. Its precise molecular structure and properties make it suitable for advanced chemical synthesis applications.

  • Functions as a drug intermediate
  • Essential for synthesizing various active compounds
  • Molecular weight of 358.11
  • Recommended storage at 4°C, protected from light
  • Solvent storage conditions include -80°C for six months or -20°C for one month, both protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.