Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Fluorophenoxy)benzyl Bromide, TCI America™
SDP

Catalog No. F0236100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0236100MG 100 mg
F02361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0236100MG Supplier TCI America Supplier No. F0236100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 242812-04-0
Molecular Formula C13H10BrFO
Molecular Weight (g/mol) 281.124
MDL Number MFCD00059914
InChI Key FWAHWPGVFDHNCH-UHFFFAOYSA-N
Synonym 3′C-Bromomethyl-2-fluorodiphenyl Ether
PubChem CID 2737454
IUPAC Name 1-(bromomethyl)-3-(2-fluorophenoxy)benzene
SMILES C1=CC=C(C(=C1)OC2=CC=CC(=C2)CBr)F

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 281.12
Physical Form Liquid
Chemical Name or Material 3-(2-Fluorophenoxy)benzyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.