Learn More
Medchemexpress LLC 3-(2-furoyl)quinoline-2-carboxaldehyde | 126769-01-5 | MFCD03427748 | 98.2% | 251.24 g/mol | C15H9NO3 | 10 MG
SDP
Catalog No. 5000297486
Encompass_Preferred
3-(2-furoyl)quinoline-2-carboxaldehyde is a fluorescent quinoline-derived aldehyde used as a fluorogenic labeling reagent in biochemical and analytical applications. It is supplied as a solid with high reported purity and is commonly used to derivatize primary amines to produce fluorescent products for HPLC, microscopy, and other assays.
  • Fluorogenic reagent for labeling primary amines.
  • High purity suitable for analytical work.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Available in small mass quantities for research use (e.g., 10 mg).
  • Stable when stored protected from light at low temperatures.
Catalog No. 50-002-97486 Supplier Medchemexpress LLC Supplier No. HYD009610MG
3-(2-furoyl)quinoline-2-carboxaldehyde is a fluorescent quinoline-derived aldehyde used as a fluorogenic labeling reagent in biochemical and analytical applications. It is supplied as a solid with high reported purity and is commonly used to derivatize primary amines to produce fluorescent products for HPLC, microscopy, and other assays.
  • Fluorogenic reagent for labeling primary amines.
  • High purity suitable for analytical work.
  • Supplied as an off-white to light yellow solid for ease of handling.
  • Available in small mass quantities for research use (e.g., 10 mg).
  • Stable when stored protected from light at low temperatures.