Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Furyl)acrolein, 99%

Catalog No. AAA1304206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304206 5 g
AAA1304214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1304206 Supplier Thermo Scientific Chemicals Supplier No. A1304206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 623-30-3 | C7H6O2 | 122.123 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-30-3
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.123
MDL Number MFCD00003256
InChI Key VZIRCHXYMBFNFD-HNQUOIGGSA-N
Synonym 3-2-furyl acrolein, 2-furanacrolein, e-3-furan-2-yl acrylaldehyde, 3-furan-2-yl acrylaldehyde, 2-propenal, 3-2-furanyl, trans-3-2-furyl acrolein, beta-2-furylacrolein, 3-2-furyl acrylaldehyde, unii-ssq86u3drp, furyl acrolein
PubChem CID 1549521
IUPAC Name (E)-3-(furan-2-yl)prop-2-enal
SMILES C1=COC(=C1)C=CC=O

Specifications

Melting Point 49°C to 52°C
Boiling Point 95°C (5 mmHg)
Flash Point 99°C (210°F)
Quantity 5 g
UN Number UN1759
Beilstein 107570
Sensitivity Air sensitive
Formula Weight 122.12
Percent Purity 99%
Chemical Name or Material 3-(2-Furyl)acrolein
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber 210-785-3
RTECSNumber LT8528500
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.