Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(2-Furyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC46104EA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC46104EA 10 g
CC46104CB 250 mg
CC46104DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC46104EA Supplier Thermo Scientific Chemicals Supplier No. CC46104EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85553-52-2
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD03990461
InChI Key BTAANNDAXIYWAN-UHFFFAOYSA-N
Synonym 3-2-furyl benzaldehyde, 3-furan-2-yl benzaldehyde, 3-furyl phenyl ketone, benzaldehyde, 3-2-furanyl, ketone, 3-furyl phenyl, 3-furan-2-yl-benzaldehyde, 3-furanylphenylmethanone, 3-furfanylphenylmethanone, 2-3-formylphenyl furan, 3-2-furanyl benzaidehyde
PubChem CID 201640
IUPAC Name 3-(furan-2-yl)benzaldehyde
SMILES O=CC1=CC=CC(=C1)C1=CC=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material 3-(2-Furyl)benzaldehyde
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.