Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-[2-hydroxy-5-(trifluoromethyl)phenyl]-6-(trifluoromethyl)-1H-benzimidazol-2-one | 153587-01-0 | MFCD00928190 | 98.1% | 362.23 g/mol | C15H8F6N2O2 | 100MG
SDP

Catalog No. 5000235182
Encompass_Preferred

NS-1619 is a small-molecule opener of large-conductance Ca2+-activated K+ (BK) channels used in pharmacology and physiology research. It is supplied as a solid or as a DMSO solution and is commonly used to probe ion channel function and vascular smooth muscle relaxation.

  • Opens large-conductance Ca2+-activated K+ (BK) channels.
  • Used to study vascular smooth muscle relaxation and ion channel physiology.
  • Reported purity: 98.1%.
  • Molecular weight: 362.23 g/mol.
  • Chemical formula: C15H8F6N2O2.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Appearance: white to off-white powder; recommended storage at -20°C for long-term stability.

Catalog No. 50-002-35182 Supplier Medchemexpress LLC Supplier No. HY12496100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NS-1619 is a small-molecule opener of large-conductance Ca2+-activated K+ (BK) channels used in pharmacology and physiology research. It is supplied as a solid or as a DMSO solution and is commonly used to probe ion channel function and vascular smooth muscle relaxation.

  • Opens large-conductance Ca2+-activated K+ (BK) channels.
  • Used to study vascular smooth muscle relaxation and ion channel physiology.
  • Reported purity: 98.1%.
  • Molecular weight: 362.23 g/mol.
  • Chemical formula: C15H8F6N2O2.
  • Available as solid powder and as 10 mM solution in DMSO.
  • Appearance: white to off-white powder; recommended storage at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.