Learn More
Medchemexpress LLC 3-[2-hydroxy-5-(trifluoromethyl)phenyl]-6-(trifluoromethyl)-1H-benzimidazol-2-one | 153587-01-0 | MFCD00928190 | 98.1% | 362.23 g·mol⁻¹ | C15H8F6N2O2 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY1249610MM 1ML
Encompass_Preferred
NS-1619 is a small-molecule opener of large-conductance Ca2+-activated potassium (BK) channels, used in electrophysiology and pharmacology research to probe BK channel function and modulation. It is supplied as a solid and as a 10 mM solution in DMSO for laboratory use, with documented solubility, storage recommendations, and physicochemical identifiers to support experimental reproducibility.
  • Opens large-conductance Ca2+-activated potassium channels for functional studies.
  • Provided as solid and 10 mM DMSO solution for flexible experimental workflows.
  • High reported purity suitable for research applications.
  • Documented solubility and in vivo formulation options for dosing studies.
  • Characterized molecular weight and chemical formula to support analytical verification.
Catalog No. 50-002-38734