Learn More
Medchemexpress LLC 3-(2-iodoethoxy)prop-1-yne | 164118-56-3 | MFCD24459258 | 95.0% | 210.01 g/mol | C5H7IO | 250 MG
SDP
Catalog No. 5000281224
Encompass_Preferred
3-(2-iodoethoxy)prop-1-yne is an alkyne-containing crosslinker used for copper-catalyzed azide-alkyne click chemistry and synthetic conjugation. The iodoether group serves as an effective leaving group for nucleophilic substitution, enabling downstream modification and bioconjugation in organic synthesis workflows.
  • Reactive terminal alkyne for efficient click chemistry conjugation.
  • Iodoether leaving group enables nucleophilic substitution.
  • Liquid physical state for easy handling and accurate dosing.
  • Suitable for bioconjugation and small-molecule synthesis applications.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-81224 Supplier Medchemexpress LLC Supplier No. HYW251756250MG
3-(2-iodoethoxy)prop-1-yne is an alkyne-containing crosslinker used for copper-catalyzed azide-alkyne click chemistry and synthetic conjugation. The iodoether group serves as an effective leaving group for nucleophilic substitution, enabling downstream modification and bioconjugation in organic synthesis workflows.
  • Reactive terminal alkyne for efficient click chemistry conjugation.
  • Iodoether leaving group enables nucleophilic substitution.
  • Liquid physical state for easy handling and accurate dosing.
  • Suitable for bioconjugation and small-molecule synthesis applications.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.