Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propionate 98.0+%, TCI America™
SDP

Catalog No. M23591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M23591G Supplier TCI America Supplier No. M23591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24249-95-4
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD28386109
InChI Key CSWRCKVZMMKVDC-UHFFFAOYSA-N
PubChem CID 57605655
IUPAC Name 3-[dimethyl({2-[(2-methylprop-2-enoyl)oxy]ethyl})azaniumyl]propanoate
SMILES CC(=C)C(=O)OCC[N+](C)(C)CCC([O-])=O

Specifications

Melting Point 101°C
Color White
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-[[2-(Methacryloyloxy)ethyl]dimethylammonio]propionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.